Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-1,5-diaminopentane 98.0+%, TCI America™
SDP

Catalog No. A13741G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13741G 1 g
A13745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A13741G Supplier TCI America Supplier No. A13741G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 51644-96-3
Molecular Formula C10H22N2O2
Molecular Weight (g/mol) 202.298
MDL Number MFCD00210020
InChI Key DPLOGSUBQDREOU-UHFFFAOYSA-N
Synonym N-(5-Aminoamyl)carbamic Acid tert-Butyl Ester, N-(5-Aminopentyl)carbamic Acid tert-Butyl Ester, N-Boc-1,5-diaminopentane, N-(tert-Butoxycarbonyl)-1,5-pentanediamine, tert-Butyl N-(5-Aminopentyl)carbamate, N-Boc-1,5-pentanediamine
PubChem CID 4352
IUPAC Name tert-butyl N-(5-aminopentyl)carbamate
SMILES CC(C)(C)OC(=O)NCCCCCN

Specifications

Color Yellow
Quantity 1 g
Formula Weight 202.30
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material N-(tert-Butoxycarbonyl)-1,5-diaminopentane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.