Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-1H-pyrazole-1-carboxamidine 98.0+%, TCI America™
SDP

Catalog No. B40285G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B40285G Supplier TCI America Supplier No. B40285G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 152120-61-1
Molecular Formula C9H14N4O2
Molecular Weight (g/mol) 210.237
MDL Number MFCD00216663
InChI Key IGSFMHYSWZUENI-UHFFFAOYSA-N
Synonym n-boc-1-guanylpyrazole, n-boc-1h-pyrazole-1-carboxamidine, n-boc-pyrazole-1-carboxamidine, n-tert-butoxycarbonyl-1h-pyrazole-1-carboxamidine, tert-butyl 1e-amino 1h-pyrazol-1-yl methylene carbamate, n-tert-butoxycarbonyl-1 h-pyrazole-1-carboxamidine, h-pyrazole-1-carboxamidine hcl, 1h-pyrazole-n-t-butyloxycarbonyl-1-carboxamidine, tert-butoxy-n-iminopyrazolylmethyl carboxamide, tert-butyl imino 1h-pyrazol-1-yl methylcarbamate
PubChem CID 9605068
IUPAC Name tert-butyl (NE)-N-[amino(pyrazol-1-yl)methylidene]carbamate
SMILES CC(C)(C)OC(=O)N=C(N)N1C=CC=N1

Specifications

Melting Point 101°C
Color White
Quantity 5 g
Formula Weight 210.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-1H-pyrazole-1-carboxamidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.