Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-2,2'-(ethylenedioxy)diethylamine 97.0+%, TCI America™
SDP

Catalog No. B5141200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5141200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B5141200MG Supplier TCI America Supplier No. B5141200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Building block for the synthesis of proteolysis targeting compounds.

Chemical Identifiers

CAS 153086-78-3
Molecular Formula C11H24N2O4
Molecular Weight (g/mol) 248.323
MDL Number MFCD03788155
InChI Key OCUICOFGFQENAS-UHFFFAOYSA-N
Synonym N-Boc-2,2′C-(ethylenedioxy)diethylamine, N-(tert-Butoxycarbonyl)-3,6-dioxa-1,8-octanediamine, N-Boc-3,6-dioxa-1,8-octanediamine, tert-Butyl [2-[2-(2-Aminoethoxy)ethoxy]ethyl]carbamate, [2-[2-(2-Aminoethoxy)ethoxy]ethyl]carbamic Acid tert-Butyl Ester
PubChem CID 9881394
IUPAC Name tert-butyl N-[2-[2-(2-aminoethoxy)ethoxy]ethyl]carbamate
SMILES CC(C)(C)OC(=O)NCCOCCOCCN

Specifications

Color Yellow
Quantity 200 mg
Formula Weight 248.32
Percent Purity ≥97.0% (HPLC,T)
Physical Form Liquid
Chemical Name or Material N-(tert-Butoxycarbonyl)-2,2′-(ethylenedioxy)diethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.