Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-3-(3-pyridyl)-L-alanine, TCI America™
SDP

Catalog No. B43245G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B43245G Supplier TCI America Supplier No. B43245G

missing translation for 'validMainframeMsg'

Specifications

Melting Point 140°C
CAS 117142-26-4
Color White
Molecular Formula C13H17N2O4
MDL Number MFCD00079681
Synonym boc-3-3-pyridyl-l-alanine, boc-3-pal-oh, boc-3-3-pyridyl-alanine, boc-l-3-3-pyridyl-alanine, s-2-tert-butoxycarbonyl amino-3-pyridin-3-yl propanoic acid, boc-3-3-pyridyl-ala-oh, boc-ala 3-pyridyl-oh, boc-l-3-pyridylala, boc-3'-pyridyl-l-ala, n-tert-butoxycarbonyl-3-3-pyridyl-l-alanine
InChI Key JLBCSWWZSSVXRQ-JTQLQIEISA-M
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=CN=C1)C([O-])=O
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(pyridin-3-yl)propanoate
Molecular Weight (g/mol) 265.29
PubChem CID 7020919
Formula Weight 266.30
Percent Purity ≥98.0% (HPLC,T)
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-3-(3-pyridyl)-L-alanine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.