Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-3-pyrroline 98.0+%, TCI America™
SDP

Catalog No. B37355G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37355G 5 g
B373525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37355G Supplier TCI America Supplier No. B37355G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 73286-70-1
Molecular Formula C9H15NO2
Molecular Weight (g/mol) 169.224
MDL Number MFCD01863512
InChI Key YEBDZDMYLQHGGZ-UHFFFAOYSA-N
Synonym tert-butyl 2,5-dihydro-1h-pyrrole-1-carboxylate, n-boc-2,5-dihydropyrrole, n-boc-3-pyrroline, 1-boc-2,5-dihydro-1h-pyrrole, tert-butyl 2h-pyrrole-1 5h-carboxylate, n-boc-2,5-dihydro-1h-pyrrole, 2,5-dihydro-pyrrole-1-carboxylic acid tert-butyl ester, 3-pyrroline, n-boc protected, 1h-pyrrole-1-carboxylic acid, 2,5-dihydro-, 1,1-dimethylethyl ester, tert-butyl-2,5-dihydro-1h-pyrrole-1-carboxylate
PubChem CID 5148195
IUPAC Name tert-butyl 2,5-dihydropyrrole-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC=CC1

Specifications

Melting Point 41°C
Boiling Point 75°C
Color White-Yellow
Quantity 5 g
Formula Weight 169.22
Percent Purity ≥98.0% (GC)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-3-pyrroline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.