Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-4-oxo-L-proline 98.0+%, TCI America™
SDP

Catalog No. B41415G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41415G 5 g
B41411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41415G Supplier TCI America Supplier No. B41415G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 84348-37-8
Molecular Formula C10H15NO5
Molecular Weight (g/mol) 229.23
MDL Number MFCD01860669
InChI Key CKYGSXRXTIKGAJ-UHFFFAOYNA-N
Synonym n-boc-4-oxo-l-proline, s-1-tert-butoxycarbonyl-4-oxopyrrolidine-2-carboxylic acid, n-t-boc-4-oxo-l-proline, boc-4-oxo-pro-oh, n-tert-boc-4-oxo-l-proline, 2s-1-tert-butoxycarbonyl-4-oxopyrrolidine-2-carboxylic acid, 1-tert-butoxycarbonyl-4-oxoproline, 2s-1-tert-butoxycarbonyl-4-oxo-pyrrolidine-2-carboxylic acid, 2s-1-tert-butoxy carbonyl-4-oxopyrrolidine-2-carboxylic acid, s-4-oxo-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester
PubChem CID 11593804
IUPAC Name 1-[(tert-butoxy)carbonyl]-4-oxopyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CC(=O)CC1C(O)=O

Specifications

Melting Point 158°C
Color White
Quantity 5 g
Formula Weight 229.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-4-oxo-L-proline
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.