Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)aminoferrocene 98.0+%, TCI America™
SDP

Catalog No. B22271G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B22271G Supplier TCI America Supplier No. B22271G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Organometallic chemical reagent intended for laboratory research purposes.

Chemical Identifiers

CAS 339195-19-6
Molecular Formula C15H19FeNO2
Molecular Weight (g/mol) 301.17
MDL Number MFCD03844774
Synonym N-Boc-aminoferrocene

Specifications

Quantity 1 g
Formula Weight 301.17
Percent Purity ≥98.0% (HPLC,T)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)aminoferrocene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.