Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-D-2-cyclohexylglycine 98.0+%, TCI America™
SDP

Catalog No. B49751G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49751G 1 g
B49755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49751G Supplier TCI America Supplier No. B49751G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 70491-05-3
Molecular Formula C13H23NO4
Molecular Weight (g/mol) 257.33
MDL Number MFCD00133629
InChI Key QSUXZIPXYDQFCX-SNVBAGLBSA-N
Synonym boc-d-chg-oh, boc-d-cyclohexylglycine, r-2-tert-butoxycarbonyl amino-2-cyclohexylacetic acid, boc-alpha-cyclohexyl-d-glycine, n-boc-2-cyclohexyl-d-glycine, 2r-n-tert-butoxycarbonyl-2-cyclohexylglycine, r-tert-butoxycarbonylamino-cyclohexyl-acetic acid, r-tert-butoxycarbonyl amino cyclohexyl acetic acid, 2r-tert-butoxycarbonyl amino cyclohexyl acetic acid, 2r-2-tert-butoxycarbonylamino-2-cyclohexyl-acetic acid
PubChem CID 7014890
IUPAC Name (2R)-2-cyclohexyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid
SMILES CC(C)(C)OC(=O)NC(C1CCCCC1)C(=O)O

Specifications

Melting Point 75°C
Color White
Quantity 1 g
Formula Weight 257.33
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-D-2-cyclohexylglycine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.