Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-D-aspartic Acid 98.0+%, TCI America™
SDP

Catalog No. B29655G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B29655G Supplier TCI America Supplier No. B29655G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 62396-48-9
Molecular Formula C9H15NO6
Molecular Weight (g/mol) 233.22
MDL Number MFCD00798618
InChI Key KAJBMCZQVSQJDE-RXMQYKEDSA-N
Synonym boc-d-aspartic acid, boc-d-asp-oh, n-boc-d-aspartic acid, n-tert-butoxycarbonyl-d-aspartic acid, d-aspartic acid, n-1,1-dimethylethoxy carbonyl, 2r-2-tert-butoxycarbonyl amino butanedioic acid, boc-d-asparticacid, pubchem12159, n-t-boc-d-aspartic acid, ksc494c3p
PubChem CID 7021110
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanedioic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)O

Specifications

Color White
Quantity 5 g
Formula Weight 233.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-D-aspartic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.