Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(tert-butoxycarbonyl)-D-glutamic acid | 34404-28-9 | MFCD00190790 | >97.0% | 247.25 g/mol | C10H17NO6 | 100 G
SDP

Catalog No. 5000301220
Encompass_Preferred

BOC-D-GLU-OH is the N-tert-butoxycarbonyl (Boc) protected D-enantiomer of glutamic acid used as a research reagent and building block in peptide synthesis and biochemical studies. The compound is supplied as a solid with manufacturer guidance for storage and documentation.

  • Boc-protected D-glutamic acid suitable for peptide synthesis and biochemical research.
  • Molecular formula C10H17NO6 and molecular weight 247.25 g/mol.
  • White to off-white powder appearance.
  • Available in multiple laboratory pack sizes (25 g, 100 g, 500 g).
  • Manufacturer storage guidance: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Purity documented by suppliers; explicit assay varies by source.

Catalog No. 50-003-01220 Supplier Medchemexpress LLC Supplier No. HYW039449100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BOC-D-GLU-OH is the N-tert-butoxycarbonyl (Boc) protected D-enantiomer of glutamic acid used as a research reagent and building block in peptide synthesis and biochemical studies. The compound is supplied as a solid with manufacturer guidance for storage and documentation.

  • Boc-protected D-glutamic acid suitable for peptide synthesis and biochemical research.
  • Molecular formula C10H17NO6 and molecular weight 247.25 g/mol.
  • White to off-white powder appearance.
  • Available in multiple laboratory pack sizes (25 g, 100 g, 500 g).
  • Manufacturer storage guidance: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Purity documented by suppliers; explicit assay varies by source.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.