Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-D-proline 98.0+%, TCI America™
SDP

Catalog No. B297725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B297725G 25 g
B29775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B297725G Supplier TCI America Supplier No. B297725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 37784-17-1
Molecular Formula C10H17NO4
Molecular Weight (g/mol) 215.249
MDL Number MFCD00063226
InChI Key ZQEBQGAAWMOMAI-SSDOTTSWSA-N
Synonym n-boc-d-proline, boc-d-proline, boc-d-pro-oh, r-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, n-tert-butoxycarbonyl-d-proline, 2r-1-tert-butoxy carbonyl pyrrolidine-2-carboxylic acid, n-t-boc-d-proline, boc-d-pro, n-alpha-t-boc-d-proline, 1-tert-butoxycarbonyl-d-proline
PubChem CID 688022
IUPAC Name (2R)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCCC1C(=O)O

Specifications

Melting Point 135°C
Color White
Quantity 25 g
Formula Weight 215.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-D-proline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.