Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-tert-butoxycarbonyl-DL-phenylglycine | 3601-66-9 | MFCD00971063 | 100.0% | 251.28 g·mol^-1 | C13H17NO4 | 100 G
SDP

Catalog No. 5000303967
Encompass_Preferred

Boc-DL-Phg-OH is a Boc-protected DL-phenylglycine derivative used as a protected amino acid building block in peptide synthesis and as a reagent in small-molecule synthesis. It is provided as a high-purity solid for research and synthetic applications.

  • Boc-protected DL-phenylglycine amino acid derivative
  • High purity suitable for synthetic chemistry
  • Molecular weight 251.28 g·mol^-1
  • Chemical formula C13H17NO4
  • Supplied as a powder; store cold to preserve stability

Catalog No. 50-003-03967 Supplier Medchemexpress LLC Supplier No. HYW013906100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-DL-Phg-OH is a Boc-protected DL-phenylglycine derivative used as a protected amino acid building block in peptide synthesis and as a reagent in small-molecule synthesis. It is provided as a high-purity solid for research and synthetic applications.

  • Boc-protected DL-phenylglycine amino acid derivative
  • High purity suitable for synthetic chemistry
  • Molecular weight 251.28 g·mol^-1
  • Chemical formula C13H17NO4
  • Supplied as a powder; store cold to preserve stability

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.