Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(tert-Butoxycarbonyl)-L-alanine-3,3,3-d3 | 161602-47-7 | MFCD00084110 | 99.0% | 192.23 | 25 MG
SDP

Catalog No. 5000402276
Encompass_Preferred

Boc-L-Ala-OH-d3 is a deuterium labeled form of Boc-L-Ala-OH, utilized as a tracer and internal standard in quantitative analysis. Its stable heavy isotopes are valuable for drug development, influencing pharmacokinetic and metabolic profiles.

  • Isotopic purity of 99 atom % D
  • Solid form
  • Optical activity of [α]20/D -23°, c = 2 in acetic acid
  • Melting point of 79-83 °C (lit.)
  • Suitable for bio NMR techniques
  • Used in peptide synthesis
  • Functional group is Boc
  • Mass shift of M+3
  • Can be used as a tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS

Catalog No. 50-004-02276 Supplier Medchemexpress LLC Supplier No. 25MGHY41121S3
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Boc-L-Ala-OH-d3 is a deuterium labeled form of Boc-L-Ala-OH, utilized as a tracer and internal standard in quantitative analysis. Its stable heavy isotopes are valuable for drug development, influencing pharmacokinetic and metabolic profiles.

  • Isotopic purity of 99 atom % D
  • Solid form
  • Optical activity of [α]20/D -23°, c = 2 in acetic acid
  • Melting point of 79-83 °C (lit.)
  • Suitable for bio NMR techniques
  • Used in peptide synthesis
  • Functional group is Boc
  • Mass shift of M+3
  • Can be used as a tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.