Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-L-alanine 98.0+%, TCI America™
SDP

Catalog No. B118425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B118425G 25 g
B11845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B118425G Supplier TCI America Supplier No. B118425G
Add to Cart
Add to Cart

N-(tert-Butoxycarbonyl)-L-alanine, >-98.0% (HPLC,T), C8H15NO4, CAS Number-15761-38-3, boc-ala-oh, boc-l-alanine, tert-butoxycarbonyl-l-alanine, boc-l-ala-oh, tert-butoxycarbonyl alanine, boc-ala, n-tert-butoxycarbonyl-l-alanine, n-t-boc-l-alanine, n-boc-l-alanine, n-alpha-t-boc-l-alanine, 25g, White

N-(tert-Butoxycarbonyl)-L-alanine, >-98.0% (HPLC,T), C8H15NO4, CAS Number-15761-38-3, boc-ala-oh, boc-l-alanine, tert-butoxycarbonyl-l-alanine, boc-l-ala-oh, tert-butoxycarbonyl alanine, boc-ala, n-tert-butoxycarbonyl-l-alanine, n-t-boc-l-alanine, n-boc-l-alanine, n-alpha-t-boc-l-alanine, 25g, White

Chemical Identifiers

CAS 15761-38-3
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.21
MDL Number MFCD00037225
InChI Key QVHJQCGUWFKTSE-YFKPBYRVSA-N
Synonym boc-ala-oh, n-tert-butoxycarbonyl-l-alanine, boc-l-alanine, n-boc-l-alanine, boc-l-ala-oh, n-t-boc-l-alanine, boc-ala, tert-butoxycarbonyl-l-alanine, tert-butoxycarbonyl alanine, n-alpha-t-boc-l-alanine
PubChem CID 85082
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
SMILES C[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 83°C
Color White
Quantity 25 g
Formula Weight 189.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-alanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.