Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-(Tert-Butoxycarbonyl)-L-Cysteine Methyl Ester

Catalog No. 4671545ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-715-45ML 5 mL
46-715-425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 46-715-45ML Supplier Sigma Aldrich Supplier No. 4671545ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-(tert-Butoxycarbonyl)-L-cysteine methyl ester is an N-terminal protected reagent that can be used in the synthesis of peptides and proteins containing cysteine residues, which can further be converted to dehydroalanine.

Specifications

CAS 55757-46-5
Density 1.143 g/mL at 25°C
Boiling Point 214°C
Molecular Formula C9H17NO4S
Linear Formula HSCH2CH[NHCO2C(CH3)3]CO2CH3
Refractive Index n20/D 1.475
MDL Number MFCD00799495
Quantity 5 mL
Synonym N-Boc-L-cysteine methyl ester
Molecular Weight (g/mol) 235.3 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.