Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-L-glutamic Acid 98.0+%, TCI America™
SDP

Catalog No. B217725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B217725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B217725G Supplier TCI America Supplier No. B217725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2419-94-5
Molecular Formula C10H15NO6
Molecular Weight (g/mol) 245.23
MDL Number MFCD00037297
InChI Key AQTUACKQXJNHFQ-LURJTMIESA-L
Synonym boc-glu-oh, boc-l-glutamic acid, n-boc-l-glutamic acid, boc-l-glu-oh, n-tert-butoxycarbonyl-l-glutamic acid, n-t-butoxycarbonyl-l-glutamic acid, n-alpha-t-boc-l-glutamic acid, n-tert-butoxycarbonyl glutamic acid, l-glutamic acid, n-1,1-dimethylethoxy carbonyl, n-alpha-t-butyloxycarbonyl-l-glutamic acid
PubChem CID 2724758
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}pentanedioate
SMILES CC(C)(C)OC(=O)N[C@@H](CCC([O-])=O)C([O-])=O

Specifications

Color White
Quantity 25 g
Formula Weight 247.25
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-glutamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.