Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-L-pyroglutamic Acid 98.0+%, TCI America™
SDP

Catalog No. B40835G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40835G 5 g
B408325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40835G Supplier TCI America Supplier No. B40835G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 53100-44-0
Molecular Formula C10H15NO5
Molecular Weight (g/mol) 229.23
MDL Number MFCD00672316
InChI Key MJLQPFJGZTYCMH-LURJTMIESA-N
Synonym boc-pyr-oh, n-boc-l-pyroglutamic acid, s-boc-5-oxopyrrolidine-2-carboxylic acid, boc-l-pyroglutamic acid, 2s-1-tert-butoxy carbonyl-5-oxopyrrolidine-2-carboxylic acid, s-1-tert-butoxycarbonyl-5-oxopyrrolidine-2-carboxylic acid, 1,2-pyrrolidinedicarboxylic acid, 5-oxo-, 1-1,1-dimethylethyl ester, 2s, 1-tert-butoxycarbonyl-5-oxo-l-proline, bocpyr, 2s-1-tert-butoxycarbonyl-5-oxopyrrolidine-2-carboxylic acid
PubChem CID 104412
IUPAC Name (2S)-1-[(tert-butoxy)carbonyl]-5-oxopyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1[C@@H](CCC1=O)C(O)=O

Specifications

Melting Point 119°C
Color White
Quantity 5 g
Formula Weight 229.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-pyroglutamic Acid
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.