Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-(Tert-Butoxycarbonyl)-L-Valine Methyl Ester

Catalog No. 4664685ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-646-85ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 46-646-85ML Supplier Sigma Aldrich Supplier No. 4664685ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-(Tert-Butoxycarbonyl)-L-Valine Methyl Ester

Specifications

CAS 70681-41-3
Density 1.232 g/mL at 25°C
Boiling Point 104°C to 108°C (1 mmHg, lit.)
Molecular Formula C6H10O5
Linear Formula CH3O2CCH2CH(OH)CO2CH3
Refractive Index n20/D 1.44
MDL Number MFCD00066216
Quantity 5 mL
Molecular Weight (g/mol) 162.14 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.