Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-N-methyl-1,2-ethylenediamine 95.0+%, TCI America™
SDP

Catalog No. B42775ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42775ML 5 mL
B42771ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42775ML Supplier TCI America Supplier No. B42775ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 121492-06-6
Molecular Formula C8H18N2O2
Molecular Weight (g/mol) 174.244
MDL Number MFCD01317789
InChI Key QYJVBVKFXDHFPQ-UHFFFAOYSA-N
Synonym n-boc-n-methylethylenediamine, n-2-aminoethyl-n-methyl carbamic acid tert-butyl ester, n-boc-2-aminoethyl-n-methylamine, tert-butyl n-2-aminoethyl-n-methylcarbamate, n-boc-n-methyl-ethylenediamine, tert-butyl 2-aminoethyl methyl carbamate, 2-n-boc-n-methylamino ethylamine, n-2-aminoethyl-n-methylcarbamicacidtert-butylester, n-boc-n-methyl ethylenediamine
PubChem CID 2756424
IUPAC Name tert-butyl N-(2-aminoethyl)-N-methylcarbamate
SMILES CC(C)(C)OC(=O)N(C)CCN

Specifications

Boiling Point 79°C
Color Colorless
UN Number 2735
Quantity 5 mL
Formula Weight 174.24
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-(tert-Butoxycarbonyl)-N-methyl-1,2-ethylenediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.