Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-N-methyl-D-phenylalanine 98.0+%, TCI America™
SDP

Catalog No. B50115G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50115G 5 g
B50111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50115G Supplier TCI America Supplier No. B50115G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 85466-66-6
Molecular Formula C15H21NO4
Molecular Weight (g/mol) 279.34
MDL Number MFCD00151890
InChI Key AJGJINVEYVTDNH-UHFFFAOYNA-N
Synonym boc-n-me-d-phe-oh, boc-d-mephe-oh, n-boc-n-methyl-d-phenylalanine, n-tert-butoxycarbonyl-n-methyl-d-phenylalanine, n-methyl-boc-d-phenylalanine, r-2-tert-butoxycarbonyl methyl amino-3-phenylpropanoic acid, 2r-2-n-tert-butoxycarbonyl-n-methylamino-3-phenylpropionic acid, boc-d-me-phe-oh, ambotzbaa1044, 2r-2-tert-butoxycarbonyl methyl amino-3-phenylpropanoic acid
PubChem CID 7015127
IUPAC Name 2-{[(tert-butoxy)carbonyl](methyl)amino}-3-phenylpropanoic acid
SMILES CN(C(CC1=CC=CC=C1)C(O)=O)C(=O)OC(C)(C)C

Specifications

Melting Point 86°C
Color White-Yellow
Quantity 5 g
Formula Weight 279.34
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-N-methyl-D-phenylalanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.