Learn More
Medchemexpress LLC N-(tert-butoxycarbonyl)-N-methylglycine | 13734-36-6 | MFCD00037795 | 98.0% | 189.21 | C8H15NO4 | 500 G
SDP
Catalog No. 5000302341
Encompass_Preferred
Boc-Sar-OH (N-(tert-butoxycarbonyl)-N-methylglycine; CAS 13734-36-6) is a Boc-protected sarcosine derivative used as a building block in peptide synthesis and amino-acid modification. Supplied as a research reagent at high purity, it is employed in peptide coupling and synthetic workflows requiring N-methylglycine residues.
  • Boc-protected N-methylglycine suitable for peptide synthesis.
  • High purity for research applications.
  • Compatible with standard peptide coupling reagents and protocols.
  • Soluble in common organic solvents such as DMSO.
  • Available in bulk quantities to support scale-up and synthesis.
Catalog No. 50-003-02341 Supplier Medchemexpress LLC Supplier No. HYW015230500G
Boc-Sar-OH (N-(tert-butoxycarbonyl)-N-methylglycine; CAS 13734-36-6) is a Boc-protected sarcosine derivative used as a building block in peptide synthesis and amino-acid modification. Supplied as a research reagent at high purity, it is employed in peptide coupling and synthetic workflows requiring N-methylglycine residues.
  • Boc-protected N-methylglycine suitable for peptide synthesis.
  • High purity for research applications.
  • Compatible with standard peptide coupling reagents and protocols.
  • Soluble in common organic solvents such as DMSO.
  • Available in bulk quantities to support scale-up and synthesis.