Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)nortropinone 98.0+%, TCI America™
SDP

Catalog No. B34011G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34011G 1 g
B34015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34011G Supplier TCI America Supplier No. B34011G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 185099-67-6
Molecular Formula C12H19NO3
Molecular Weight (g/mol) 225.288
MDL Number MFCD00673779
InChI Key MENILFUADYEXNU-UHFFFAOYSA-N
Synonym tert-butyl 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylate, n-boc-nortropinone, n-boc-4-nortropinone, n-tert-butoxycarbonyl nortropinone, 8-boc-8-azabicyclo 3.2.1 octan-3-one, 3-oxo-8-aza-bicyclo 3.2.1 octane-8-carboxylic acid tert-butyl ester, t-butyl 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylate, 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylic acid tert-butyl ester, n-boc-nororopinone, 8-tert-butoxycarbonyl-8-azabicyclo 3.2.1 octan-3-one;
PubChem CID 2794767
IUPAC Name tert-butyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate
SMILES CC(C)(C)OC(=O)N1C2CCC1CC(=O)C2

Specifications

Melting Point 71°C
Color White-Yellow
Quantity 1 g
Formula Weight 225.29
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)nortropinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.