Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyloxy)phthalimide 95.0+%, TCI America™
SDP

Catalog No. C15735G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C15735G Supplier TCI America Supplier No. C15735G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15263-20-4
Molecular Formula C13H13NO5
Molecular Weight (g/mol) 263.249
MDL Number MFCD00005889
InChI Key MCWXBNWFVFOQAS-UHFFFAOYSA-N
Synonym tert-Butyl Phthalimido Carbonate, Carbonic Acid tert-Butyl Phthalimido Ester
PubChem CID 84857
IUPAC Name tert-butyl (1,3-dioxoisoindol-2-yl) carbonate
SMILES CC(C)(C)OC(=O)ON1C(=O)C2=CC=CC=C2C1=O

Specifications

Color White
Quantity 5 g
Formula Weight 263.25
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyloxy)phthalimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.