Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-tert-Butyl-3-methylpyridine-2-carboxamide

Catalog No. KOB00291G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
KOB00291G 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. KOB00291G Supplier Sigma Aldrich Supplier No. KOB00291G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-methoxypyridine has been prepared from 4-methoxypyridine-n-oxide, via catalytic hydrogenation
  • Ortho lithiation of 4-methoxypyridine using mesityllithium as the metalating base has been studied.4-methoxypyridine was used as a starting reagent for the stereocontrolled synthesis of (+/-)-pumiliotoxin c and (+/-)-lasubine ii
  • It was also used in an efficient construction of dihyropyridin-4-ones, which serves as potential ligands for neuronal nicotinic acetycholine receptors

Specifications

CAS 32998-95-1
Molecular Formula C11H16N2O
Linear Formula C11H16N2O
MDL Number MFCD04108091
Quantity 5 mL
Molecular Weight (g/mol) 192.26
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.