Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-tert-Butyl-4-fluorobenzamide, 97%, Thermo Scientific™

Catalog No. AAH57752MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH57752MD 250 mg
AAH5775203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH57752MD Supplier Thermo Scientific Chemicals Supplier No. H57752MD

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 49834-29-9
Molecular Formula C11H14FNO
Molecular Weight (g/mol) 195.237
MDL Number MFCD00463095
InChI Key ZMYTVKMZSNODPU-UHFFFAOYSA-N
Synonym n-t-butyl-4-fluorobenzamide, n-tert-butyl-4-fluorobenzamide, n-tert-butyl-4-fluoro-benzamide, acmc-209kih
PubChem CID 4295826
IUPAC Name N-tert-butyl-4-fluorobenzamide
SMILES CC(C)(C)NC(=O)C1=CC=C(C=C1)F

Specifications

Quantity 250 mg
Formula Weight 195.24
Percent Purity 97%
Chemical Name or Material N-tert-Butyl-4-fluorobenzamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.