Learn More
AdipoGen N-tert-Butyl-alpha-phenylnitro
SDP
Catalog No. 502561612
Encompass_Preferred
Chemical. CAS 3376-24-7. Formula C11H15NO. MW 177.2. Synthetic. Cell permeable spin trap commonly used in free radical research for both in vivo and in vitro studies. It protects against oxidative damage caused by various inflammatory events, demonstrating neuroprotective, anti-anging, and antidiabetic effects. PBN has been shown to inhibit LPS-induced NF-kappaB DNA binding activity, inhibit COX-2 catalytic activity, inhibit lipid peroxidation in rat liver microsomes and prevent the induction of inducible nitric oxide synthase iNOS. Shows anti-cancer activity in several experimental cancer models.
Catalog No. 50-256-1612 Supplier AdipoGen Supplier No. CDXB0269G005
Chemical. CAS 3376-24-7. Formula C11H15NO. MW 177.2. Synthetic. Cell permeable spin trap commonly used in free radical research for both in vivo and in vitro studies. It protects against oxidative damage caused by various inflammatory events, demonstrating neuroprotective, anti-anging, and antidiabetic effects. PBN has been shown to inhibit LPS-induced NF-kappaB DNA binding activity, inhibit COX-2 catalytic activity, inhibit lipid peroxidation in rat liver microsomes and prevent the induction of inducible nitric oxide synthase iNOS. Shows anti-cancer activity in several experimental cancer models.