Learn More
Abcam N-tert-Butyl-alpha-phenylnitrone (PBN), 500MG
Catalog No. 502704488
Encompass_Preferred
MW 177.24 Da, Purity >98%. Cell permeable free radicle trap. Inhibits LPS-induced NF-κB DNA binding activity, inhibits COX-2 catalytic activity and inhibits lipid peroxidation in rat liver microsomes. Blocks indiction of iNOS by reactive oxygen species.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4488 Supplier Abcam Supplier No. AB146814500MG
MW 177.24 Da, Purity >98%. Cell permeable free radicle trap. Inhibits LPS-induced NF-κB DNA binding activity, inhibits COX-2 catalytic activity and inhibits lipid peroxidation in rat liver microsomes. Blocks indiction of iNOS by reactive oxygen species.
The product is subject to the following: Abcam Restricted Use Statement