Learn More
Medchemexpress LLC N-tert-butyl-n-(2-chlorophenyl)-3-(isoquinolinyloxy)propanamide | 142720-24-9 | MFCD00467704 | 100.0% | 436.60 g/mol | C28H37FN2O | 25 MG
SDP
Catalog No. 5000212921
Encompass_Preferred
FGIN 1-27 is an indoleacetamide research compound that functions as a high-affinity agonist of the translocator protein (TSPO), used in neuroscience and pharmacology studies to probe TSPO-mediated mechanisms. It is supplied for laboratory research in solid and solution formats and is suitable for in vitro and preclinical assays.
  • High-affinity TSPO agonist with reported Ki ≈ 5 nM.
  • High purity suitable for research applications.
  • Available in multiple formats, including solid vials and 10 mM solution in DMSO.
  • Molecular weight 436.60 g/mol and formula C28H37FN2O for compound identification.
  • Offered in small-scale quantities suitable for assay development and dose-ranging studies.
Catalog No. 50-002-12921 Supplier Medchemexpress LLC Supplier No. HY10105925MG
FGIN 1-27 is an indoleacetamide research compound that functions as a high-affinity agonist of the translocator protein (TSPO), used in neuroscience and pharmacology studies to probe TSPO-mediated mechanisms. It is supplied for laboratory research in solid and solution formats and is suitable for in vitro and preclinical assays.
  • High-affinity TSPO agonist with reported Ki ≈ 5 nM.
  • High purity suitable for research applications.
  • Available in multiple formats, including solid vials and 10 mM solution in DMSO.
  • Molecular weight 436.60 g/mol and formula C28H37FN2O for compound identification.
  • Offered in small-scale quantities suitable for assay development and dose-ranging studies.