Learn More
Sigma Aldrich Fine Chemicals Biosciences N tert Butyldimethylsilyl
Catalog No. 502897934
Encompass
N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide is a silylating reagent which replaces the active hydrogen with tert-Butyldimethylsilyl group. The tert-Butyldimethylsilyl derivatives are found to be more resistant to hydrolysis and stable compared to trimethylsilyl (TMS) derivatives. This reagent is suitable for GC-MS analysis since it produces mass spectra which can be easily interpreted. It is widely used for the silylation of different alcohols thiols phenols carboxylic acids amines and amides.
Catalog No. 50-289-7934 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 7762610X1ML
N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide is a silylating reagent which replaces the active hydrogen with tert-Butyldimethylsilyl group. The tert-Butyldimethylsilyl derivatives are found to be more resistant to hydrolysis and stable compared to trimethylsilyl (TMS) derivatives. This reagent is suitable for GC-MS analysis since it produces mass spectra which can be easily interpreted. It is widely used for the silylation of different alcohols thiols phenols carboxylic acids amines and amides.