Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Tert-Butyldimethylsilyl-N-Methyltrifluoroacetamide

Catalog No. 3948821ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
10 x 1 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-488-21ML 10 x 1 mL
39-488-25ML 5 mL
39-488-2100ML 100 mL
39-488-225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Warning
Catalog No. 39-488-21ML Supplier Sigma Aldrich Supplier No. 39488210X1ML
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-tert-butyldimethylsilyl-N-methyltrifluoroacetamide (MTBSTFA) can be used as an efficient silylating agent for different alcohols, thiols, phenols, carboxylic acids, amines, and amides. It can also be used to functionalize multi-walled carbon nanotubes (MWCNTs) with polar groups. MTBSTFA is reportedly much less moisture-sensitive than other silylating reagents used for GLC derivatization.

Specifications

CAS 77377-52-7
Density 1.036 g/mL at 25°C
Boiling Point 172°C to 175°C
Molecular Formula C9H18F3NOSi
Refractive Index n20/D 1.402
Linear Formula CF3CON(CH3)Si(CH3)2C(CH3)3
MDL Number MFCD00009671
Quantity 10 x 1 mL
Synonym MTBSTFA
Molecular Weight (g/mol) 241.33 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.