Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Tert-Butyldimethylsilyl-N-Methyltrifluoroacetamide With 1% Tert-Butyldimethylchlorosilane

Catalog No. 37593425ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
10 x 1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-593-425ML 25 mL
37-593-45ML 5 mL
37-593-41ML 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Warning
Catalog No. 37-593-425ML Supplier Sigma Aldrich Supplier No. 37593425ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide with 1% tert-butyldimethylchlorosilane is commonly used for the preparation of N-tert-butyldimethylsilyl ethanolamines resulting from the hydrolysis of nitrogen mustards. It is also used for selective O-silylation of N,O-diacylhydroxyl amines.

Specifications

CAS 77377-52-7
Molecular Formula C9H18F3NOSi
Linear Formula C9H18F3NOSi
MDL Number MFCD00009671
Quantity 25 mL
Synonym MTBSTFA+TBDMSCl, 99:1; N-Methyl-N-tert-butyldimethylsilyltrifluoroacetamide; MTBSTFA + 1% TBDMSCl; Silylating mixture VI
Molecular Weight (g/mol) 241.33 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.