Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-tert-Butylmaleimide 98.0+%, TCI America™
SDP

Catalog No. B36385G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36385G 5 g
B36381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36385G Supplier TCI America Supplier No. B36385G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4144-22-3
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.181
MDL Number MFCD00191885
InChI Key YEKDUBMGZZTUDY-UHFFFAOYSA-N
PubChem CID 643190
IUPAC Name 1-tert-butylpyrrole-2,5-dione
SMILES CC(C)(C)N1C(=O)C=CC1=O

Specifications

Color Yellow
Boiling Point 190°C
Quantity 5 g
Formula Weight 153.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-tert-Butylmaleimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.