Learn More
Medchemexpress LLC N-tetradecanoyl-DL-homoserine lactone | 98206-80-5 | MFCD01862913 | 97.2% | 311.46 g/mol | C18H33NO3 | 10 MG
SDP
Catalog No. 5000282889
Encompass_Preferred
N-Tetradecanoyl-DL-homoserine lactone is an N-acyl homoserine lactone research reagent used to study bacterial quorum sensing and cell-to-cell signaling. Provided as a powder with an accompanying certificate of analysis, the compound is chemically defined (C18H33NO3, 311.46 g/mol) and intended for in vitro research applications.
  • High purity: 97.2% (LCMS) as supplied on COA.
  • Identified by CAS number 98206-80-5.
  • Powder form suitable for laboratory handling and formulation.
  • Recommended storage: sealed, away from moisture; powder -80 °C (2 years) or -20 °C (1 year).
  • Supplied with a certificate of analysis and lot-specific data.
Catalog No. 50-002-82889 Supplier Medchemexpress LLC Supplier No. HYW14209310MG
N-Tetradecanoyl-DL-homoserine lactone is an N-acyl homoserine lactone research reagent used to study bacterial quorum sensing and cell-to-cell signaling. Provided as a powder with an accompanying certificate of analysis, the compound is chemically defined (C18H33NO3, 311.46 g/mol) and intended for in vitro research applications.
  • High purity: 97.2% (LCMS) as supplied on COA.
  • Identified by CAS number 98206-80-5.
  • Powder form suitable for laboratory handling and formulation.
  • Recommended storage: sealed, away from moisture; powder -80 °C (2 years) or -20 °C (1 year).
  • Supplied with a certificate of analysis and lot-specific data.