Learn More
Strem, An Ascensus Company CAS# 4671-75-4. 1g. n-Tetradecylphosphonic acid, min. 97% TDPA. MFCD00015834
SDP
Catalog No. 5090110305
Encompass
CAS# 4671-75-4. 1g. n-Tetradecylphosphonic acid, min. 97% TDPA. MFCD00015834. Molecular Weight: 278.37. Molecular Formula: CH3(CH2)13P(O)(OH)2. Color/form: white to off-white pwdr. Strem# 15-5145. http://www.strem.com/catalog/v/15-5145/
Catalog No. 50-901-10305 Supplier Strem, An Ascensus Company Supplier No. 1551451G
CAS# 4671-75-4. 1g. n-Tetradecylphosphonic acid, min. 97% TDPA. MFCD00015834. Molecular Weight: 278.37. Molecular Formula: CH3(CH2)13P(O)(OH)2. Color/form: white to off-white pwdr. Strem# 15-5145. http://www.strem.com/catalog/v/15-5145/