Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Organic Chemistry N,N,N',N'-Tetramethyl-1,4-butanediamine | 5G | 111-51-3 | MFCD00008338 | 0.98

Catalog No. 1271085G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred

N,N,N',N'-Tetramethyl-1,4-butanediamine, 5G

About This Item:

Density: 0.792 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.428 (lit.)
Storage: room temp
EINECS Number: 203-878-5

Catalog No. 12-710-85G Supplier Sigma Organic Chemistry Supplier No. 1271085G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N,N,N',N'-Tetramethyl-1,4-butanediamine, 5G

About This Item:

Density: 0.792 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.428 (lit.)
Storage: room temp
EINECS Number: 203-878-5

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.