Learn More
Crescent Chemical Co Inc N-TETRATRIACONTANE SOLN 1ML
SDP
Supplier:  Crescent Chemical Co Inc CT3853D
Encompass_Preferred
n-Tetratriacontane Single Component Calibration Standard in Dichloromethane CAS 14167-59-0 Size - 1mL Catalog Number - 3853D
Catalog No. 50-696-62