Learn More
Medchemexpress LLC N-trans-Caffeoyltyramine | 103188-48-3 | 1 MG
SDP
Catalog No. 5000417479
Encompass_Preferred
N-trans-Caffeoyltyramine is an effective inflammatory response regulator with antioxidant activity and anticoagulation effects. It is intended for research use only and has not been validated for medical applications.
  • Effective inflammatory response regulator
  • Antioxidant activity
  • Anticoagulation effects
  • Acts on human SH-SY5Y (SH) cells
  • Changes microRNA expression spectrum of nerve cells
Catalog No. 50-004-17479 Supplier Medchemexpress LLC Supplier No. HYN82411MG
N-trans-Caffeoyltyramine is an effective inflammatory response regulator with antioxidant activity and anticoagulation effects. It is intended for research use only and has not been validated for medical applications.
  • Effective inflammatory response regulator
  • Antioxidant activity
  • Anticoagulation effects
  • Acts on human SH-SY5Y (SH) cells
  • Changes microRNA expression spectrum of nerve cells