Learn More
Medchemexpress LLC N-trans-Caffeoyltyramine | 103188-48-3 | 1 ML
SDP
Catalog No. 5000417417
Encompass_Preferred
N-trans-Caffeoyltyramine is an effective inflammatory response regulator that exhibits antioxidant and anticoagulation effects. It is intended for research use only.
  • Targets reactive oxygen species (ROS)
  • Involved in immunology/inflammation and NF-kB pathways
  • Acts on human SH-SY5Y cells with an IC50 value of 59 μM
  • Alters the microrna expression spectrum of nerve cells, lowering MIR-199A-5P by 50%
Catalog No. 50-004-17417 Supplier Medchemexpress LLC Supplier No. HYN824110MM/1ML
N-trans-Caffeoyltyramine is an effective inflammatory response regulator that exhibits antioxidant and anticoagulation effects. It is intended for research use only.
  • Targets reactive oxygen species (ROS)
  • Involved in immunology/inflammation and NF-kB pathways
  • Acts on human SH-SY5Y cells with an IC50 value of 59 μM
  • Alters the microrna expression spectrum of nerve cells, lowering MIR-199A-5P by 50%