Learn More
Medchemexpress LLC N-trans-Caffeoyltyramine | 103188-48-3 | 25 MG
SDP
Catalog No. 5000421889
Encompass_Preferred
N-trans-Caffeoyltyramine is an effective inflammatory response regulator, possessing antioxidant activity and anticoagulation effects. This product is intended for research use only.
  • Regulates inflammatory response
  • Has antioxidant activity
  • Exhibits anticoagulation effects
  • Targets reactive oxygen species (ROS)
  • Acts on human SH-SY5Y cells with an IC50 value of 59 μM
  • Can alter the microrna expression spectrum of nerve cells
Catalog No. 50-004-21889 Supplier Medchemexpress LLC Supplier No. 25MGHYN8241
N-trans-Caffeoyltyramine is an effective inflammatory response regulator, possessing antioxidant activity and anticoagulation effects. This product is intended for research use only.
  • Regulates inflammatory response
  • Has antioxidant activity
  • Exhibits anticoagulation effects
  • Targets reactive oxygen species (ROS)
  • Acts on human SH-SY5Y cells with an IC50 value of 59 μM
  • Can alter the microrna expression spectrum of nerve cells