Learn More
Medchemexpress LLC N-trans-Caffeoyltyramine | 103188-48-3 | MFCD24849356 | 5 MG
SDP
Catalog No. 5000419148
Encompass_Preferred
N-trans-Caffeoyltyramine is an effective inflammatory response regulator, which has antioxidant activity and anticoagulation effects. It is intended for research use only and possesses a high purity of 99.72%.
  • Effective inflammatory response regulator.
  • Possesses antioxidant activity.
  • Has anticoagulation effects.
  • Lowers MIR-199A-5P by 50% in human SH-SY5Y (SH) cells.
  • Appears as a white to off-white solid.
Catalog No. 50-004-19148 Supplier Medchemexpress LLC Supplier No. HYN82415MG
N-trans-Caffeoyltyramine is an effective inflammatory response regulator, which has antioxidant activity and anticoagulation effects. It is intended for research use only and possesses a high purity of 99.72%.
  • Effective inflammatory response regulator.
  • Possesses antioxidant activity.
  • Has anticoagulation effects.
  • Lowers MIR-199A-5P by 50% in human SH-SY5Y (SH) cells.
  • Appears as a white to off-white solid.