Learn More
TARGETMOL CHEMICALS INC N-TRANS-FERULOYLTYRAMINE 5MG
SDP
Catalog No. 502580016
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. N-trans-Feruloyltyramine (Feruloyltyramine)(NTF) has hepatoprotective effect. 2. NTF has antioxidative activity against A(beta)(1-42)-induced neuronal death. 3. NTF is likely to inhibit COX enzymes thereby suppressing P-selectin expression on platelets is a platelet aggregation inhibitor. 4. NTF inhibits melanogenesis in a dose-dependent manner NTF induces downregulation of tyrosinase resulted in suppression of melanin biosynthesis in murine B16 melanoma cells. purity: 99%
Catalog No. 50-258-0016 Supplier TARGETMOL CHEMICALS INC Supplier No. T3S06455MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. N-trans-Feruloyltyramine (Feruloyltyramine)(NTF) has hepatoprotective effect. 2. NTF has antioxidative activity against A(beta)(1-42)-induced neuronal death. 3. NTF is likely to inhibit COX enzymes thereby suppressing P-selectin expression on platelets is a platelet aggregation inhibitor. 4. NTF inhibits melanogenesis in a dose-dependent manner NTF induces downregulation of tyrosinase resulted in suppression of melanin biosynthesis in murine B16 melanoma cells. purity: 99%