Learn More
Medchemexpress LLC N-trans-Feruloyltyramine | 66648-43-9 | 1 MG
SDP
Catalog No. 5000421133
Encompass_Preferred
N-trans-Feruloyltyramine is an alkaloid and a potent antioxidant. It improves H2O2-induced intracellular Reactive Oxygen Species (ROS) generation and decreases apoptosis. It has potential for research in oxidative stress-related neurodegenerative conditions and cancer.
  • Protects cells against H2O2-induced toxicity.
  • Significantly reduces Bax and activated caspase-3 levels in H2O2-treated SK-N-SH cells.
  • Significantly improves an H2O2-mediated increase in ROS levels.
  • Does not affect viability of SK-N-SH cells.
  • Shows proliferation inhibition on HepG2 cells.
Catalog No. 50-004-21133 Supplier Medchemexpress LLC Supplier No. HYN24101MG
N-trans-Feruloyltyramine is an alkaloid and a potent antioxidant. It improves H2O2-induced intracellular Reactive Oxygen Species (ROS) generation and decreases apoptosis. It has potential for research in oxidative stress-related neurodegenerative conditions and cancer.
  • Protects cells against H2O2-induced toxicity.
  • Significantly reduces Bax and activated caspase-3 levels in H2O2-treated SK-N-SH cells.
  • Significantly improves an H2O2-mediated increase in ROS levels.
  • Does not affect viability of SK-N-SH cells.
  • Shows proliferation inhibition on HepG2 cells.