Learn More
Apexbio Technology LLC N-trans-Feruloyltyramine 66648-43-9 1mg
SDP
Catalog No. 5000089099
Encompass_Preferred
N-trans-Feruloyltyramine (CAS 66648-43-9) is a naturally occurring alkaloid recognized for its antioxidant properties Mechanistically it reduces intracellular reactive oxygen species (ROS) levels and mitigates apoptosis in response to oxidative agents such as hydrogen peroxide Through its modulation of oxidative stress pathways N-trans-Feruloyltyramine is of interest in studies exploring the pathogenesis and intervention of neurodegenerative disorders and cancer associated with oxidative damage This compound thus serves as a useful tool for investigating oxidative stress and its cellular effects in biomedical research
Catalog No. 50-000-89099 Supplier Apexbio Technology LLC Supplier No. BA29751
N-trans-Feruloyltyramine (CAS 66648-43-9) is a naturally occurring alkaloid recognized for its antioxidant properties Mechanistically it reduces intracellular reactive oxygen species (ROS) levels and mitigates apoptosis in response to oxidative agents such as hydrogen peroxide Through its modulation of oxidative stress pathways N-trans-Feruloyltyramine is of interest in studies exploring the pathogenesis and intervention of neurodegenerative disorders and cancer associated with oxidative damage This compound thus serves as a useful tool for investigating oxidative stress and its cellular effects in biomedical research