Learn More
Medchemexpress LLC N-trans-Feruloyltyramine | 66648-43-9 | 3.13 MG
SDP
Catalog No. 5000421125
Encompass_Preferred
N-trans-Feruloyltyramine, an alkaloid, acts as a potent antioxidant. It has demonstrated the ability to improve H2O2-induced intracellular Reactive Oxygen Species (ROS) generation and decrease apoptosis. This compound holds potential for research into oxidative stress-related neurodegenerative conditions and cancer.
  • Protects cells against H2O2-induced toxicity
  • Significantly reduces Bax and activated caspase-3 levels in H2O2-treated SK-N-SH cells
  • Significantly improves H2O2-mediated increase in ROS levels
Catalog No. 50-004-21125 Supplier Medchemexpress LLC Supplier No. HYN241010MM/1ML
N-trans-Feruloyltyramine, an alkaloid, acts as a potent antioxidant. It has demonstrated the ability to improve H2O2-induced intracellular Reactive Oxygen Species (ROS) generation and decrease apoptosis. This compound holds potential for research into oxidative stress-related neurodegenerative conditions and cancer.
  • Protects cells against H2O2-induced toxicity
  • Significantly reduces Bax and activated caspase-3 levels in H2O2-treated SK-N-SH cells
  • Significantly improves H2O2-mediated increase in ROS levels