Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-trans-p-coumaroyloctopamine | 66648-45-1 | MFCD22417271 | 99.7% | 299.32 | C17H17NO4 | 10 MG
SDP

Catalog No. 5000296301
Encompass_Preferred

N-trans-p-coumaroyloctopamine is a phenylpropanoid amide isolated from eggplant (Solanum melongena L.) and provided for research use. It is offered as a high-purity solid and as a convenient DMSO solution, suitable as a reference compound in natural product and bioactivity studies.

  • High purity (99.7%).
  • Molecular formula C17H17NO4 and molecular weight 299.32.
  • CAS number 66648-45-1 for unambiguous identification.
  • Available in lab-scale solid quantities and solution formats for convenient handling.
  • Intended for chemical and biological research applications.

Catalog No. 50-002-96301 Supplier Medchemexpress LLC Supplier No. HYN223110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-trans-p-coumaroyloctopamine is a phenylpropanoid amide isolated from eggplant (Solanum melongena L.) and provided for research use. It is offered as a high-purity solid and as a convenient DMSO solution, suitable as a reference compound in natural product and bioactivity studies.

  • High purity (99.7%).
  • Molecular formula C17H17NO4 and molecular weight 299.32.
  • CAS number 66648-45-1 for unambiguous identification.
  • Available in lab-scale solid quantities and solution formats for convenient handling.
  • Intended for chemical and biological research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.