Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Tridecylamine 98%, Thermo Scientific™

Catalog No. AC139550250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC139550250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC139550250 Supplier Thermo Scientific Chemicals Supplier No. 139550250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2869-34-3
Molecular Formula C13H29N
Molecular Weight (g/mol) 199.382
InChI Key ABVVEAHYODGCLZ-UHFFFAOYSA-N
Synonym 1-aminotridecane, 1-tridecanamine, tridecylamine, n-tridecylamine, monotridecylamine, tridecanamine, 1-tridecylamine, unii-5s0kq9wz2d, 5s0kq9wz2d, tridecyl-amine
PubChem CID 40761
IUPAC Name tridecan-1-amine
SMILES CCCCCCCCCCCCCN

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 26°C to 30°C
Color Colorless to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula CH3(CH2)12NH2
Quantity 25 g
Beilstein 04, 201
Formula Weight 199.38
Percent Purity ≥97.5%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material n-Tridecylamine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.