Learn More
Medchemexpress LLC N-trimethoxysilylpropyl-N,N,N-trimethylammonium chloride | 35141-36-7 | MFCD00054225 | 50% in methanol (solution) | C9H24ClNO3Si | 500 MG
SDP
Catalog No. 5000433298
Encompass_Preferred
N-trimethoxysilylpropyl-N,N,N-trimethylammonium chloride, supplied as a 50% solution in methanol, is a biochemical reagent. It is suitable for use as a biological material or organic compound in life science research and is intended for research use only.
  • Suitable for life science research
  • Supplied as a 50% solution in methanol
  • Liquid appearance, colorless to light yellow
  • Store at -20°C, protected from light
  • Stable for 2 years when stored correctly
  • For research use only
Catalog No. 50-004-33298 Supplier Medchemexpress LLC Supplier No. HY91018500MG
N-trimethoxysilylpropyl-N,N,N-trimethylammonium chloride, supplied as a 50% solution in methanol, is a biochemical reagent. It is suitable for use as a biological material or organic compound in life science research and is intended for research use only.
  • Suitable for life science research
  • Supplied as a 50% solution in methanol
  • Liquid appearance, colorless to light yellow
  • Store at -20°C, protected from light
  • Stable for 2 years when stored correctly
  • For research use only