Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(Trimethylsilyl)dimethylamine 95.0+%, TCI America™
SDP

Catalog No. T059125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T059125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T059125ML Supplier TCI America Supplier No. T059125ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2083-91-2
Molecular Formula C5H15NSi
Molecular Weight (g/mol) 117.27
MDL Number MFCD00008297
InChI Key KAHVZNKZQFSBFW-UHFFFAOYSA-N
Synonym n,n-dimethyltrimethylsilylamine, n,n-dimethylaminotrimethylsilane, silanamine, pentamethyl, dimethylamino trimethylsilane, pentamethylsilylamine, n-trimethylsilyl dimethylamine, tmsdma, dimethylaminotrimethylsilane, silanamine, n,n,1,1,1-pentamethyl, n,n-dimethyltrimethylsilamine
PubChem CID 74965
IUPAC Name dimethyl(trimethylsilyl)amine
SMILES CN(C)[Si](C)(C)C

Specifications

Color Yellow
Boiling Point 86°C
Quantity 25 mL
UN Number 2733
Formula Weight 117.27
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-(Trimethylsilyl)dimethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.