Learn More
Cayman Chemical N-trans-Feruloyltyramine, Purity: ≥98%, CAS: 66648-43-9, 5 mg
SDP
Catalog No. 502941616
Encompass_Preferred
A phenolic amide with diverse biological activities; decreases NO production (IC50 = 17.36 µM) and iNOS activity and increases NO scavenging without inducing cytotoxicity in LPS-stimulated BV-2 cells; scavenges DPPH radicals in cell-free assays; inhibits proliferation of HeLa and L929 cells by 72.2 and 22%, respectively, at 30 µg/ml
Catalog No. 50-294-1616 Supplier Cayman Chemical Supplier No. 274595 MG
A phenolic amide with diverse biological activities; decreases NO production (IC50 = 17.36 µM) and iNOS activity and increases NO scavenging without inducing cytotoxicity in LPS-stimulated BV-2 cells; scavenges DPPH radicals in cell-free assays; inhibits proliferation of HeLa and L929 cells by 72.2 and 22%, respectively, at 30 µg/ml