Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Undecylamine 98%, Thermo Scientific™

Catalog No. AC140700500
Change view
Click to view available options
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC140700500 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC140700500 Supplier Thermo Scientific Chemicals Supplier No. 140700500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7307-55-3
CAS Min % 97.5
CAS Max % 100.0
Density 0.796g/mL
Assay Percent Range 98%
Molecular Formula C11H25N
Linear Formula CH3(CH2)10NH2
Quantity 50mL
MDL Number MFCD00008152
Beilstein 04, 199
Synonym undecylamine, n-undecylamine, 1-aminoundecane, 1-undecanamine, hendecylamine, 1-undecylamine, monoundecylamine, n-undecylamin, unii-uze26a4ko1, uze26a4ko1
InChI Key QFKMMXYLAPZKIB-UHFFFAOYSA-N
SMILES CCCCCCCCCCCN
IUPAC Name undecan-1-amine
Molecular Weight (g/mol) 171.33
PubChem CID 81742
Formula Weight 171.33
Chemical Name or Material n-Undecylamine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 230-761-6
RTECSNumber YQ1518000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.